Learn More
SOMNI Scientific™ SOMNI Scientific™ SOMNI Ped Coaxial Circuit
SDP
Catalog No. 04777208 Shop All Somni Scientific Products
Change view
Click to view available options
Length (English):
40 in.
60 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
04-777-208 40 in.
04-777-207 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-777-208 Supplier SOMNI Scientific™ Supplier No. BC13013
Add to Cart
Add to Cart
The tube within a tube design eliminates twisting of the inspiratory and expiratory limbs and provides heat and moisture conservation. PVC, Phthalate and Latex Free
  • A tube within a tube design that eliminates twisting of the inspiratory and expiratory limbs.
  • PVC, Phthalate and Latex Free
  • Provides heat and moisture conservation
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Length (English) 40 in.