Learn More
SOMNI Scientific™ SOMNI Scientific™ Stopcock Assembly
SDP
Catalog No. 04777196 Shop All Somni Scientific Products
Change view
Click to view available options
Diameter (English):
0.25 in.
Catalog No. Diameter (English)
04-777-196 0.25 in.
1 options
Catalog No. 04-777-196 Supplier SOMNI Scientific™ Supplier No. AS8002
Add to Cart
Add to Cart
Open the stopcock to allow gas flow or close it when not in use. Both ends fit snugly into the supply tubing.
  • The Stopcock assembly is a convenient device to incorporate into supply tubing
Specifications
Diameter (English) 0.25 in.
For Use With (Application) Research Animal Inhalant Anesthesia