Learn More
SOMNI Scientific™ SOMNI Scientific™ TEC 5 Iso KF Vaporizer
SDP
Catalog No. 04777194 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-194 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-194 Supplier SOMNI Scientific™ Supplier No. VS6019
Add to Cart
Add to Cart
Refurbished vaporizer that includes a cagemount with hose barbs. This eliminates the need for 23 mm adapters at the inlet and outlet and the possibility of leaks. High liquid capacity that means less refilling.
  • The Tec 5 Isoflurane key fill is a refurbished vaporizer that includes a cagemount with hose barbs
  • hose barb cagemount eliminates the need for 23 mm adapters
  • Reduced possibility of leaks
  • High liquid capacity that means less refilling
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia