Learn More
SOMNI Scientific™ SOMNI Scientific™ TT2 Anesthesia Syst
SDP
Catalog No. 04777214 Shop All Somni Scientific Products
Change view
Click to view available options
Includes:
SOMNI 3 extended capacity vaporizer
Catalog No. Includes
04-777-214 SOMNI 3 extended capacity vaporizer
1 options
Catalog No. 04-777-214 Supplier SOMNI Scientific™ Supplier No. TT17001
Add to Cart
Add to Cart
Request a Quote
Allows the user to provide anesthesia through one or both diverter valves without disconnecting supply lines
  • Allows the user to provide anesthesia through one or both diverter valves without disconnecting supply lines
  • TT-2 tabletop anesthesia system (TT-17000)
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Includes SOMNI 3 extended capacity vaporizer