Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonepiprazole, Tocris Bioscience™

Catalog No. 418510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-851-0 10 mg
41-855-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-851-0 Supplier R&D Systems Supplier No. 4185/10
This item is not returnable. View return policy

Selective D4 receptor antagonist

Selective, high affinity antagonist for the rat and human dopamine D4 receptor (Ki = 10 nM). Exhibits low affinity at other monoamine receptors (Ki > 2000 nM). Induces c-fos gene expression in medial prefrontal cortex in a similar manner to clozapine (Cat. No. 0444). Brain penetrant and orally available.

Chemical Identifiers

CAS 170858-33-0
Molecular Formula C21H27N3O3S
Molecular Weight (g/mol) 401.525
InChI Key WNUQCGWXPNGORO-NRFANRHFSA-N
Synonym sonepiprazole, isochr-etpip-phso2nh2, sonepiprazole hydrate, chembl69759, 4-4-2-isochroman-1-yl ethyl piperazin-1-yl benzenesulfonamide, 4-4-2-1s-3,4-dihydro-1h-2-benzopyran-1-yl ethyl-1-piperazinyl-benzenesulfonamide, 4-4-2-1s-3,4-dihydro-1h-isochromen-1-yl ethyl piperazin-1-yl benzenesulfonamide, 4-4-2-1s-isochroman-1-yl ethyl piperazin-1-yl benzenesulfonamide hydrate, sonepiprazole inn, d01rae
PubChem CID 133079
IUPAC Name 4-[4-[2-[(1S)-3,4-dihydro-1H-isochromen-1-yl]ethyl]piperazin-1-yl]benzenesulfonamide
SMILES C1COC(C2=CC=CC=C21)CCN3CCN(CC3)C4=CC=C(C=C4)S(=O)(=O)N

Specifications

Quantity 10 mg
Formula Weight 401.52
Percent Purity >98%
Chemical Name or Material Sonepiprazole
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.