Learn More
Active Motif Sonication Tubes / Caps, 1.5 ml polystyrene
SDP
Supplier:  Active Motif 53072
Encompass
Because polystyrene transfers sonic wave energy much more efficiently than polypropylene, Active Motif highly recommends that its Polystyrene Sonication Tubes be used with the Q8000R Sonicator. Use of polypropylene tubes in indirect sonication has been shown to reduce the extent to which the chromatin is sheared.
Catalog No. NC1852598