Learn More
Sonics & Materials Inc SONICATOR 130WATT MODEL VCX130
SDP
Catalog No. NC9252207
Encompass_Preferred
VCX130 Ultrasonic Processor; 130W; Energy Monitor; Digital Wattmeter; Ten Hour Timer; 1 to 59 Second Independent ON/OFF Pulser; Variable Power Output Control; Remote Process Capability; Microprocessor Based
This item is not returnable. View return policy
Catalog No. NC9252207 Supplier Sonics & Materials Inc Supplier No. VCX130
VCX130 Ultrasonic Processor; 130W; Energy Monitor; Digital Wattmeter; Ten Hour Timer; 1 to 59 Second Independent ON/OFF Pulser; Variable Power Output Control; Remote Process Capability; Microprocessor Based