Learn More
ACROBiosystems ACROBiosystems Human Sonic Hedgehog / Shh Protein, His Tag
Catalog No. 501956865
Encompass_Preferred
Recombinant Protein;50UG;Human Sonic Hedgehog, His Tag (SH7-H5229) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Gly 197 (Accession # NP_000184).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.4 kDa. The protein migrates as 22 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-6865 Supplier ACROBiosystems Supplier No. SH7H522950UG
Recombinant Protein;50UG;Human Sonic Hedgehog, His Tag (SH7-H5229) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Gly 197 (Accession # NP_000184).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.4 kDa. The protein migrates as 22 kDa under reducing (R) condition (SDS-PAGE).