Learn More
Medchemexpress LLC HY-16582A 10mg Medchemexpress, Sonidegib CAS:956697-53-3 Purity:>98%
SDP
Catalog No. 502029205
Encompass_Preferred
Medchemexpress, HY-16582A 10mg Sonidegib CAS:956697-53-3 Sonidegib (Erismodegib) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9205 Supplier Medchemexpress LLC Supplier No. HY16582A10MG
Medchemexpress, HY-16582A 10mg Sonidegib CAS:956697-53-3 Sonidegib (Erismodegib) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.