Learn More
Medchemexpress LLC Sonidegib diphosphate | 1218778-77-8 | 99.97% | 681.49 | 10 MM 1 ML
SDP
Catalog No. 5000391796
Encompass_Preferred
Sonidegib diphosphate is a potent and selective Smoothened (Smo) antagonist, inhibiting the Hedgehog (Hh) pathway. It has demonstrated antitumor activity in medulloblastoma models and can reduce chronic myeloid leukemia (CML) stem cells.
  • Potent and selective Smo antagonist
  • Inhibits Hedgehog pathway signaling
  • Reduces expression of downstream signaling targets
  • Demonstrates dose-related antitumor activity
  • Reduces chronic myeloid leukemia stem cells
  • Associated with numerous clinical trials
Catalog No. 50-003-91796 Supplier Medchemexpress LLC Supplier No. HY1658210MM/1ML
Sonidegib diphosphate is a potent and selective Smoothened (Smo) antagonist, inhibiting the Hedgehog (Hh) pathway. It has demonstrated antitumor activity in medulloblastoma models and can reduce chronic myeloid leukemia (CML) stem cells.
  • Potent and selective Smo antagonist
  • Inhibits Hedgehog pathway signaling
  • Reduces expression of downstream signaling targets
  • Demonstrates dose-related antitumor activity
  • Reduces chronic myeloid leukemia stem cells
  • Associated with numerous clinical trials