Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sonidegib diphosphate | 1218778-77-8 | MFCD16627994 | 100.0% | 681.49 g·mol^-1 | C26H32F3N3O11P2 | 10 MG
SDP

Catalog No. 5000289270
Encompass_Preferred

Sonidegib diphosphate is the diphosphate salt of sonidegib, a potent and selective Smoothened (Smo) antagonist used in research to inhibit Hedgehog pathway signaling. It shows low-nanomolar binding potency in Smo assays and is supplied for laboratory use with supporting datasheet and safety documents.

  • Potent Smo antagonist with low-nanomolar binding (mouse 1.3 nM; human 2.5 nM).
  • Diphosphate salt form suitable for aqueous assay conditions.
  • Supplied as a 10 mg vial for research use.
  • Molecular weight 681.49 g·mol^-1; formula C26H32F3N3O11P2.
  • Storage: sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-89270 Supplier Medchemexpress LLC Supplier No. HY1658210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sonidegib diphosphate is the diphosphate salt of sonidegib, a potent and selective Smoothened (Smo) antagonist used in research to inhibit Hedgehog pathway signaling. It shows low-nanomolar binding potency in Smo assays and is supplied for laboratory use with supporting datasheet and safety documents.

  • Potent Smo antagonist with low-nanomolar binding (mouse 1.3 nM; human 2.5 nM).
  • Diphosphate salt form suitable for aqueous assay conditions.
  • Supplied as a 10 mg vial for research use.
  • Molecular weight 681.49 g·mol^-1; formula C26H32F3N3O11P2.
  • Storage: sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.