Learn More
TARGETMOL CHEMICALS INC Sonidegib diphosphate 50MG
SDP
Catalog No. 502876377
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Sonidegib diphosphate (LDE225 diphosphate) is a selective antagonist of Smo that inhibits murine Smo (IC501.3 nM) and human Smo (IC502.5 nM). Purity 99.86%
Catalog No. 50-287-6377 Supplier TARGETMOL CHEMICALS INC Supplier No. T1572750MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Sonidegib diphosphate (LDE225 diphosphate) is a selective antagonist of Smo that inhibits murine Smo (IC501.3 nM) and human Smo (IC502.5 nM). Purity 99.86%