Learn More
Enzo Life Sciences Sonidegib (free base) (100mg). CAS: 956697-53-3
SDP
Supplier:  Enzo Life Sciences ENZCHM2380100
Encompass_Preferred
Sonidegib, also known as LDE-225, is a potent and selective smoothened (Smo) antagonist. Alternative name: LDE225 (free base). Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3667