Learn More
Medchemexpress LLC Sonidegib metabolite 100mg | 1221722-10-6 | 296.24 g/mol | C15H11F3O3 | 100 MG
SDP
Catalog No. 5000228793
Encompass_Preferred
Sonidegib metabolite M48 is the primary circulating metabolite of the hedgehog-pathway inhibitor sonidegib (LDE225). Supplied as a research-grade reference material, it is used for ADME profiling, metabolite identification, and pharmacokinetic studies.
  • Primary circulating metabolite of sonidegib (M48).
  • Suitable for ADME, metabolite identification, and pharmacokinetic studies.
  • High purity (typically 98.0% by HPLC).
  • Chemical formula C15H11F3O3; molecular weight 296.24 g/mol.
  • Available in small milligram quantities for laboratory research.
Catalog No. 50-002-28793 Supplier Medchemexpress LLC Supplier No. HY117229100MG
Sonidegib metabolite M48 is the primary circulating metabolite of the hedgehog-pathway inhibitor sonidegib (LDE225). Supplied as a research-grade reference material, it is used for ADME profiling, metabolite identification, and pharmacokinetic studies.
  • Primary circulating metabolite of sonidegib (M48).
  • Suitable for ADME, metabolite identification, and pharmacokinetic studies.
  • High purity (typically 98.0% by HPLC).
  • Chemical formula C15H11F3O3; molecular weight 296.24 g/mol.
  • Available in small milligram quantities for laboratory research.