Learn More
Medchemexpress LLC Sonidegib metabolite 25mg | 1221722-10-6 | 25 MG
SDP
Catalog No. 5000226778
Encompass_Preferred
Sonidegib metabolite M48 is the primary circulating metabolite of sonidegib, provided as a research-grade chemical for laboratory studies. The compound has formula C15H11F3O3, molecular weight 296.24 g·mol⁻¹, and is supplied with documented purity and storage recommendations for research use only.
  • Research use only; not for human or clinical use.
  • High reported purity: 98.0%.
  • White to off-white solid suitable for analytical and in vitro studies.
  • Available in small research quantities for assay development and metabolic studies.
  • Stable at room temperature for up to 3 years under recommended storage conditions.
Catalog No. 50-002-26778 Supplier Medchemexpress LLC Supplier No. HY11722925MG
Sonidegib metabolite M48 is the primary circulating metabolite of sonidegib, provided as a research-grade chemical for laboratory studies. The compound has formula C15H11F3O3, molecular weight 296.24 g·mol⁻¹, and is supplied with documented purity and storage recommendations for research use only.
  • Research use only; not for human or clinical use.
  • High reported purity: 98.0%.
  • White to off-white solid suitable for analytical and in vitro studies.
  • Available in small research quantities for assay development and metabolic studies.
  • Stable at room temperature for up to 3 years under recommended storage conditions.