Learn More
Medchemexpress LLC Sonidegib metabolite 50mg | 1221722-10-6 | 296.24 g·mol⁻¹ | C15H11F3O3 | 50 MG
SDP
Catalog No. 5000226361
Encompass_Preferred
Sonidegib metabolite M48 (2-methyl-3-[4-(trifluoromethoxy)phenyl]benzoic acid) is the primary circulating metabolite of sonidegib, supplied as a research-grade reference standard for pharmacokinetic, metabolic, and analytical studies.
  • For research use only; not for human or clinical use.
  • High purity: 98.0%.
  • White to off-white solid, stable under recommended storage.
  • Comprehensive analytical data available (COA, HNMR, LC-MS).
  • Recommended storage: room temperature (solid) for up to 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Suitable as a reference standard for PK and metabolism studies.
Catalog No. 50-002-26361 Supplier Medchemexpress LLC Supplier No. HY11722950MG
Sonidegib metabolite M48 (2-methyl-3-[4-(trifluoromethoxy)phenyl]benzoic acid) is the primary circulating metabolite of sonidegib, supplied as a research-grade reference standard for pharmacokinetic, metabolic, and analytical studies.
  • For research use only; not for human or clinical use.
  • High purity: 98.0%.
  • White to off-white solid, stable under recommended storage.
  • Comprehensive analytical data available (COA, HNMR, LC-MS).
  • Recommended storage: room temperature (solid) for up to 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Suitable as a reference standard for PK and metabolism studies.