Learn More
BRANSON ULTRASONICS CORPORATION (3P) SONIFIER SFX550 20KHZ 3/4 HORN
Catalog No. NC3721546
Encompass_Preferred
Branson Ultrasonics Sonifier SFX250/SFX550 Cell Disruptors, Model: SFX550, Includes: Power supply, 20kHz converter, 1.9cm (0.75 in.) tapped horn, power cord, manual, wrench set, Wattage: 550w, Voltage: 120V, Dimensions: 1.8 x 19.1 x 23.5cm (12.5 x 7.5 x 9.25 in.)
Catalog No. NC3721546 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 101063968R
Add to Cart
Add to Cart
Request a Quote
Branson Ultrasonics Sonifier SFX250/SFX550 Cell Disruptors, Model: SFX550, Includes: Power supply, 20kHz converter, 1.9cm (0.75 in.) tapped horn, power cord, manual, wrench set, Wattage: 550w, Voltage: 120V, Dimensions: 1.8 x 19.1 x 23.5cm (12.5 x 7.5 x 9.25 in.)