Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sonlicromanol | 1541170-75-5 | 99.8% | 10 MG
SDP

Catalog No. 5000230489
Encompass_Preferred

Sonlicromanol is an orally active reactive oxygen species (ROS) modulator used for mitochondrial disease research. It is provided as a high-purity research reagent with good solubility in DMSO.

  • Orally active ROS modulator suitable for mitochondrial disease research.
  • High purity, 99.75%.
  • Molecular formula C19H28N2O3; molecular weight 332.44 g·mol⁻¹.
  • Soluble in DMSO at approximately 25 mg/mL; sonication may be required.
  • Packaged as 10 mg for laboratory research use.

Catalog No. 50-002-30489 Supplier Medchemexpress LLC Supplier No. HY12157710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sonlicromanol is an orally active reactive oxygen species (ROS) modulator used for mitochondrial disease research. It is provided as a high-purity research reagent with good solubility in DMSO.

  • Orally active ROS modulator suitable for mitochondrial disease research.
  • High purity, 99.75%.
  • Molecular formula C19H28N2O3; molecular weight 332.44 g·mol⁻¹.
  • Soluble in DMSO at approximately 25 mg/mL; sonication may be required.
  • Packaged as 10 mg for laboratory research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.