Learn More
Marketlab SONO Disinfectant Wipes
Catalog No. 22505344
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
22-505-344 50/Pk.
1 options
Catalog No. 22-505-344 Supplier Marketlab Supplier No. 17297
Add to Cart
Add to Cart
SONO Ultrasound Wipes that kill 47 pathogens and are safe for cables, lenses, and touchscreens.
  • Flat pack: 7 in. x 10 in., 50/pk.
  • Kills 47 pathogens including MRSA, HIV, Staphylococcus, and the 2013 influenza A virus H7N9.
  • Wont damage cables or lenses.
  • Safe to handle without gloves.
  • Safe on touchscreens and system consoles.
Specifications
Quantity 50/Pk.