Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonoco™ ThermoSafe LabMailer 10-Slide Shipper
GSA_VA

Catalog No. 03526 Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Description:
Holds 10 slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
03-526 Holds 10 slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-526 Supplier Sonoco™ Supplier No. 369
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maximum breakage protection for nonhazardous blood, serum, tissue, and other specimens

  • Individual slots hold up to ten 3 × 1 in. (7.6 × 2.5cm) microscope slides, with or without cover glasses
  • Specimen surfaces never touch each other
  • Plug-type lid
  • Polystyrene-foam wall thickness, 1/2 in. (13mm)

Order Info

Outside, 4L x 2.375W x 2 in. H (10 x 6 x 5cm) Inside, 2.75L x 1.5W x 1 in. D (7 x 3.8 x 2.5cm) Weight empty, 0.25 oz. (7.1g); Sleeves sold separately (03-526A)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Holds 10 slides
Dimensions (L x W x H) 4 x 2.5 x 2 in.
Material Polystyrene Foam
Quantity 10 Slides
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.