Description
- The LabMailer aluminum mailing can is designed for the secure transport of specimens.
- Material: Aluminum
- Application: Suitable for shipping frozen, refrigerated, and ambient specimens for compliance, analysis, testing, research and development, public health, industrial hygiene, and environmental use
Specifications
Specifications
| Quantity | Samples below 0 degrees C for approximately 3 hours |
| Closure Type | Lined Screw Cap |
| Material | Aluminum |