Learn More
Cayman Chemical Sonepiprazole, Purity: ≥98%, CAS: 170858-33-0, 5 mg
SDP
Catalog No. 502953164
Encompass_Preferred
A selective D4 receptor antagonist (Kis = 10 nM); selective over D1, D2, and D3 receptors, serotonin 1A and 2 receptors, as well as α1- and α2-adrenergic receptors (Kis > 2,000 nM)
Catalog No. 50-295-3164 Supplier Cayman Chemical Supplier No. 119835 MG
A selective D4 receptor antagonist (Kis = 10 nM); selective over D1, D2, and D3 receptors, serotonin 1A and 2 receptors, as well as α1- and α2-adrenergic receptors (Kis > 2,000 nM)