Learn More
Cadmet Inc SONY HIGH GLOSS PAPER 10/BOX
SDP
Catalog No. NC9744055
Encompass_Preferred
High density glossy thermal black and white printing paper for the 895MD/D895/897MD/D897 printers. Approximately 240 printers per roll. One case = 10 rolls. Prints are about 25% glossier than the UPP-110HD media - 110mm wide x 18m long.
Catalog No. NC9744055 Supplier Cadmet Inc Supplier No. 10124
High density glossy thermal black and white printing paper for the 895MD/D895/897MD/D897 printers. Approximately 240 printers per roll. One case = 10 rolls. Prints are about 25% glossier than the UPP-110HD media - 110mm wide x 18m long.