Learn More
Sony Biotechnology Sony MA900 Cell Sorter with 488nm/405nm/561nm/638nm Laser and 96 and 384 well plate deposition system
Catalog No. NC3885109
Encompass_Preferred
The MA900 Cell Sorter is designed for flexibility, enabling the detection of up to 14 parameters (12 fluorescence and 2 scatter), supporting sorting of a wide range of applications using 70um, 100um and 130um microfluidics chips into 4-way tubes. Software tools enable precision single cell sorting into multi-well plates - 96 and 384 wells and index analysis. Its design integrates advanced technologies including a patented microfluidic chip design, precise fluidic controls, and intelligent automation to streamline operation, reduce subjectivity, and enhance reliability. Patented CoreFinder technology ensures robust daily operation by automating QC, sort setup, sort monitoring and rapid clog recovery.
MA900 Multi Application Cell Sorter Brochure
Catalog No. NC3885109 Supplier Sony Biotechnology Supplier No. LEMA900FP
This product is not available for purchase online. Please contact your sales representative for a quote or further ordering instructions.
The MA900 Cell Sorter is designed for flexibility, enabling the detection of up to 14 parameters (12 fluorescence and 2 scatter), supporting sorting of a wide range of applications using 70um, 100um and 130um microfluidics chips into 4-way tubes. Software tools enable precision single cell sorting into multi-well plates - 96 and 384 wells and index analysis. Its design integrates advanced technologies including a patented microfluidic chip design, precise fluidic controls, and intelligent automation to streamline operation, reduce subjectivity, and enhance reliability. Patented CoreFinder technology ensures robust daily operation by automating QC, sort setup, sort monitoring and rapid clog recovery.
MA900 Multi Application Cell Sorter Brochure