Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab Sony Print Media Paper

Catalog No. 22030419
Change view
Click to view available options
Length (English) Page:
59.06 ft.
65.62 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Page
22-030-419 65.62 ft.
22-030-418 59.06 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-030-419 Supplier Marketlab Supplier No. 900811
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The most popular and reliable medical imaging paper

  • Temperature control and fast rotation assure fresh inventory
  • Assures clear crisp images
  • Cone Instruments assures great pricing and quick delivery on this staple supply

Specifications

Material Paper
Ruled No
Width (English) Page 4.33 in.
Length (English) Page 65.62 ft.
Color White
Dimensions (L x W) No
Product Type Printer Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.