Learn More
ABS Imaging Systems SONY THERMO PAPER UPP110HG 5PK
SDP
Catalog No. NC9275430
Encompass_Preferred
High density, high glossy thermal print paper for the UP-895MD, UP-D895MD, UP-897MD, UP-D897MD printers. Prints are about 25% glossier than the UPP-110HD media. Approximately 270 prints per roll, 5 rolls per pack.
Catalog No. NC9275430 Supplier ABS Imaging Systems Supplier No. UPP110HG
High density, high glossy thermal print paper for the UP-895MD, UP-D895MD, UP-897MD, UP-D897MD printers. Prints are about 25% glossier than the UPP-110HD media. Approximately 270 prints per roll, 5 rolls per pack.