Learn More
ABS Imaging Systems Sony UPC-R81MD Print Pack
SDP
Catalog No. NC0801781
Encompass_Preferred
US letter size dye sublimation color printing pack for use with Sony UP-DR80MD digital color printer. Self-laminating, curl free media for high print durability and longevity. Protective laminated coating is resistant to light, heat, water, humidity and fingerprints. 1 pack contains 2 ribbons and 2 rolls of media, to yield 100 prints per pack. Media length per roll 216 mm x 15 m (8 1/2 x 590 inches).
Catalog No. NC0801781 Supplier ABS Imaging Systems Supplier No. UPCR81MD
US letter size dye sublimation color printing pack for use with Sony UP-DR80MD digital color printer. Self-laminating, curl free media for high print durability and longevity. Protective laminated coating is resistant to light, heat, water, humidity and fingerprints. 1 pack contains 2 ribbons and 2 rolls of media, to yield 100 prints per pack. Media length per roll 216 mm x 15 m (8 1/2 x 590 inches).