Learn More
Selleck Chemical LLC SOP1812 E1982-25MG
SDP
Catalog No. 5000039209
Encompass_Preferred
SOP1812( QN-302) is a tetra-substituted naphthalene diimide (ND) derivative and selective inhibitor of G-quadruplex (G4) transcription It shows great affinity to hTERT G4 and HuTel21 G4 with KD values of 4 9 and 28 4 nM respectively It binds to G-quadruplex DNA in cancer gene promoters region and shows strong anti-proliferative and anti-tumor activity in pancreatic cancer models
Catalog No. 50-000-39209 Supplier Selleck Chemical LLC Supplier No. E198225MG
SOP1812( QN-302) is a tetra-substituted naphthalene diimide (ND) derivative and selective inhibitor of G-quadruplex (G4) transcription It shows great affinity to hTERT G4 and HuTel21 G4 with KD values of 4 9 and 28 4 nM respectively It binds to G-quadruplex DNA in cancer gene promoters region and shows strong anti-proliferative and anti-tumor activity in pancreatic cancer models