Learn More
Cayman Chemical Sophocarpine, Purity: ≥98%, CAS: 6483-15-4, 10 mg
SDP
Catalog No. 502988662
Encompass_Preferred
An alkaloid with diverse biological activities; cytotoxic to various cancer cell lines (IC50s = 1.64-12.86 mM); nematocidal against B. xylophilus in vitro; reduces infarct size, cardiac muscle fiber swelling and hypertrophy, and neutrophil infiltration in a rat model of myocardial ischemia and reperfusion injury at 30 mg/kg; decreases liver weight, fibrosis, and levels of IL-6, TNF-α, and TGF-β1 in a rat model of NASH; reduces carrageenan-induced hind paw edema in rats and decreases acetic acid-induced writhing in mice at 15 and 30 mg/kg
Catalog No. 50-298-8662 Supplier Cayman Chemical Supplier No. 2959810 MG
An alkaloid with diverse biological activities; cytotoxic to various cancer cell lines (IC50s = 1.64-12.86 mM); nematocidal against B. xylophilus in vitro; reduces infarct size, cardiac muscle fiber swelling and hypertrophy, and neutrophil infiltration in a rat model of myocardial ischemia and reperfusion injury at 30 mg/kg; decreases liver weight, fibrosis, and levels of IL-6, TNF-α, and TGF-β1 in a rat model of NASH; reduces carrageenan-induced hind paw edema in rats and decreases acetic acid-induced writhing in mice at 15 and 30 mg/kg