Learn More
Medchemexpress LLC Sophocarpine monohydrate | 145572-44-7 | MFCD 27923119 | 99.9% | 264.36 | C15H24N2O2 | 10 MG
SDP
Catalog No. 5000295773
Encompass_Preferred
Sophocarpine monohydrate is a naturally occurring quinolizidine alkaloid used in preclinical research. It functions as a PTEN activator and inhibits PI3K/Akt, MEK/ERK, and NF-κB signaling pathways, with reported anti-tumor, anti-inflammatory, antioxidant, and anti-apoptotic effects. The compound is supplied as a solid monohydrate, with characterized purity, solubility, and storage information for research use.
  • Activates PTEN and modulates PI3K/Akt, MEK/ERK, and NF-κB signaling pathways.
  • Shows anti-tumor and anti-inflammatory activity in preclinical models.
  • High reported purity (99.9%).
  • Supplied as a solid monohydrate suitable for formulation and storage per recommended conditions.
  • Soluble in DMSO at high concentrations; documented in vivo formulation options are available.
Catalog No. 50-002-95773 Supplier Medchemexpress LLC Supplier No. HYN0103A10MG
Sophocarpine monohydrate is a naturally occurring quinolizidine alkaloid used in preclinical research. It functions as a PTEN activator and inhibits PI3K/Akt, MEK/ERK, and NF-κB signaling pathways, with reported anti-tumor, anti-inflammatory, antioxidant, and anti-apoptotic effects. The compound is supplied as a solid monohydrate, with characterized purity, solubility, and storage information for research use.
  • Activates PTEN and modulates PI3K/Akt, MEK/ERK, and NF-κB signaling pathways.
  • Shows anti-tumor and anti-inflammatory activity in preclinical models.
  • High reported purity (99.9%).
  • Supplied as a solid monohydrate suitable for formulation and storage per recommended conditions.
  • Soluble in DMSO at high concentrations; documented in vivo formulation options are available.