Learn More
Medchemexpress LLC Sophoraisoflavone A (Allolicoisoflavone B) | 117204-81-6 | 352.34 | 1 MG
SDP
Catalog No. 5000414580
Encompass_Preferred
Sophoraisoflavone A (Allolicoisoflavone B) is a natural product isolated from Glycyrrhiza inflata. It acts as a potent protein tyrosine phosphatase 1B (PTP1B) inhibitor with an IC50 value of 0.80 μM and can be utilized in inflammation research.
  • Potent protein tyrosine phosphatase 1B (PTP1B) inhibitor.
  • IC50 value of 0.80 μM.
  • Can be used in research of inflammation.
Catalog No. 50-004-14580 Supplier Medchemexpress LLC Supplier No. HYN12301MG
Sophoraisoflavone A (Allolicoisoflavone B) is a natural product isolated from Glycyrrhiza inflata. It acts as a potent protein tyrosine phosphatase 1B (PTP1B) inhibitor with an IC50 value of 0.80 μM and can be utilized in inflammation research.
  • Potent protein tyrosine phosphatase 1B (PTP1B) inhibitor.
  • IC50 value of 0.80 μM.
  • Can be used in research of inflammation.