Learn More
Medchemexpress LLC Sophoricoside | 152-95-4 | 432.38 | 1 ML
SDP
Catalog No. 5000389973
Encompass_Preferred
Sophoricoside is an isoflavone glycoside isolated from *Sophora japonica* with anti-inflammatory, anti-cancer, and immunosuppressive effects. It significantly inhibits histamine release and reduces the levels of TNF-α, IL-8, and IL-6. It also reduces scratching behaviors in vivo and aids in the recovery of atopic dermatitis.
  • Inhibits PMACI-induced histamine release
  • Reduces TNF-α, IL-8, and IL-6 levels
  • Inhibits Rel/p65 nuclear levels
  • Reduces caspase-1 activity
  • Decreases scratching behaviors in vivo
  • Aids in recovery from atopic dermatitis
Catalog No. 50-003-89973 Supplier Medchemexpress LLC Supplier No. HYN042310MM/1ML
Sophoricoside is an isoflavone glycoside isolated from *Sophora japonica* with anti-inflammatory, anti-cancer, and immunosuppressive effects. It significantly inhibits histamine release and reduces the levels of TNF-α, IL-8, and IL-6. It also reduces scratching behaviors in vivo and aids in the recovery of atopic dermatitis.
  • Inhibits PMACI-induced histamine release
  • Reduces TNF-α, IL-8, and IL-6 levels
  • Inhibits Rel/p65 nuclear levels
  • Reduces caspase-1 activity
  • Decreases scratching behaviors in vivo
  • Aids in recovery from atopic dermatitis