Learn More
Medchemexpress LLC Sophoricoside | 152-95-4 | 432.38 | 250 MG
SDP
Catalog No. 5000389606
Encompass_Preferred
Sophoricoside is an isoflavone glycoside isolated from *Sophora japonica*. It has anti-inflammatory, anti-cancer, and immunosuppressive effects.
  • Anti-inflammatory effects
  • Anti-cancer effects
  • Immunosuppressive effects
  • Isolated from *Sophora japonica*
Catalog No. 50-003-89606 Supplier Medchemexpress LLC Supplier No. 250MGHYN0423
Sophoricoside is an isoflavone glycoside isolated from *Sophora japonica*. It has anti-inflammatory, anti-cancer, and immunosuppressive effects.
  • Anti-inflammatory effects
  • Anti-cancer effects
  • Immunosuppressive effects
  • Isolated from *Sophora japonica*