Learn More
Medchemexpress LLC Sophoricoside | 152-95-4 | 432.38 | 500 MG
SDP
Catalog No. 5000390197
Encompass_Preferred
Sophoricoside is an isoflavone glycoside isolated from Sophora japonica with anti-inflammatory, anti-cancer, and immunosuppressive effects. It has been shown to significantly inhibit histamine release and reduce levels of inflammatory cytokines like TNF-α, IL-8, and IL-6 in vitro. In vivo, it can reduce scratching behaviors and recover atopic dermatitis in mice.
  • Inhibits PMACI-induced histamine release.
  • Reduces TNF-α, IL-8, and IL-6 levels.
  • Inhibits Rel/p65 levels in the nucleus.
  • Reduces enhanced caspase-1 activity.
  • Decreases scratching behaviors.
  • Recovers atopic dermatitis in mice.
Catalog No. 50-003-90197 Supplier Medchemexpress LLC Supplier No. HYN0423500MG
Sophoricoside is an isoflavone glycoside isolated from Sophora japonica with anti-inflammatory, anti-cancer, and immunosuppressive effects. It has been shown to significantly inhibit histamine release and reduce levels of inflammatory cytokines like TNF-α, IL-8, and IL-6 in vitro. In vivo, it can reduce scratching behaviors and recover atopic dermatitis in mice.
  • Inhibits PMACI-induced histamine release.
  • Reduces TNF-α, IL-8, and IL-6 levels.
  • Inhibits Rel/p65 levels in the nucleus.
  • Reduces enhanced caspase-1 activity.
  • Decreases scratching behaviors.
  • Recovers atopic dermatitis in mice.