Learn More
Medchemexpress LLC Sophoricoside | 152-95-4 | 99.6% | 10 MG
SDP
Catalog No. 5000296261
Encompass_Preferred
Sophoricoside is an isoflavone glycoside isolated from Sophora japonica and supplied for research use. It has reported anti-inflammatory, anti-cancer, and immunosuppressive activities. The compound has molecular formula C21H20O10, molecular weight 432.38 g/mol, and CAS number 152-95-4. It is provided as a small solid pack (10 mg) with high reported purity (~99.6%).
  • Isoflavone glycoside isolated from Sophora japonica.
  • Reported anti-inflammatory, anti-cancer, and immunosuppressive effects.
  • High reported purity, suitable for analytical and biological research.
  • Molecular formula C21H20O10; molecular weight 432.38 g/mol.
  • Supplied in small solid packs for laboratory research.
Catalog No. 50-002-96261 Supplier Medchemexpress LLC Supplier No. HYN042310MG
Sophoricoside is an isoflavone glycoside isolated from Sophora japonica and supplied for research use. It has reported anti-inflammatory, anti-cancer, and immunosuppressive activities. The compound has molecular formula C21H20O10, molecular weight 432.38 g/mol, and CAS number 152-95-4. It is provided as a small solid pack (10 mg) with high reported purity (~99.6%).
  • Isoflavone glycoside isolated from Sophora japonica.
  • Reported anti-inflammatory, anti-cancer, and immunosuppressive effects.
  • High reported purity, suitable for analytical and biological research.
  • Molecular formula C21H20O10; molecular weight 432.38 g/mol.
  • Supplied in small solid packs for laboratory research.