Learn More
Medchemexpress LLC HY-N1373 5mg Medchemexpress, Sophoridine CAS:6882-68-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN13735MG
Encompass_Preferred
Medchemexpress, HY-N1373 5mg Sophoridine CAS:6882-68-4 Sophoridine is a quinolizidine alkaloid isolated from leafs of Leguminous plant Sophora alopecuroides.L. Sophoridine induces apoptosis. Sophoridine has the potential to be a novel, potent and selective antitumor drug candidate for pancreatic cancer with well-tolerated toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0331