Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sophorose monohydrate | 140686-17-5 | 99.9% | 1 MG
SDP

Catalog No. 5000234508
Encompass_Preferred

Sophorose monohydrate is a chemically defined disaccharide (CAS 140686-17-5) supplied as a solid for research applications. The material is provided in small weighed packs and is accompanied by analytical documentation, including a certificate of analysis and safety data sheet.

  • Purity: 99.9%.
  • Molecular weight: 360.31 g/mol.
  • Chemical formula: C12H24O12.
  • Physical form: solid, white to off-white.
  • Available small pack sizes, including 1 mg and 5 mg.
  • Supplied with COA and SDS documentation.

Catalog No. 50-002-34508 Supplier Medchemexpress LLC Supplier No. HY119445A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sophorose monohydrate is a chemically defined disaccharide (CAS 140686-17-5) supplied as a solid for research applications. The material is provided in small weighed packs and is accompanied by analytical documentation, including a certificate of analysis and safety data sheet.

  • Purity: 99.9%.
  • Molecular weight: 360.31 g/mol.
  • Chemical formula: C12H24O12.
  • Physical form: solid, white to off-white.
  • Available small pack sizes, including 1 mg and 5 mg.
  • Supplied with COA and SDS documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.