Learn More
Sigma Aldrich Fine Chemicals Biosciences sopropyl alcohol meets US500ML
Catalog No. 502897642
Encompass
Isopropyl alcohol (2-Propanol i-PrOH) a renewable secondary alcohol is water soluble colorless volatile liquid with a sweet odor. It is a potential substitute to petroleum-derived gasoline. The preparation of methyl isobutyl ketone (MIBK) using i-PrOH as the precursor has been reported with yield as high as 25%. The solubility of benzoic acid has been investigated in i-PrOH by gravimetric method in the temperature range 277-346K. Its photocatalytic degradation has been examined by various methods. This product is a high quality pharmacopoeia product that meets the testing specifications of USP (United States Pharmacopoeia). It can be employed in research and pilot studies.
Catalog No. 50-289-7642 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I9030500ML
Isopropyl alcohol (2-Propanol i-PrOH) a renewable secondary alcohol is water soluble colorless volatile liquid with a sweet odor. It is a potential substitute to petroleum-derived gasoline. The preparation of methyl isobutyl ketone (MIBK) using i-PrOH as the precursor has been reported with yield as high as 25%. The solubility of benzoic acid has been investigated in i-PrOH by gravimetric method in the temperature range 277-346K. Its photocatalytic degradation has been examined by various methods. This product is a high quality pharmacopoeia product that meets the testing specifications of USP (United States Pharmacopoeia). It can be employed in research and pilot studies.