Learn More
Medchemexpress LLC SOR-C13 | 1187852-48-7 | 99.1% | 1565.81 | 10 MG
SDP
Catalog No. 5000422646
Encompass_Preferred
SOR-C13 is a carboxy-terminal truncated peptide that acts as a high-affinity TRPV6 antagonist with an IC50 value of 14 nM. It exhibits anticancer activity by targeting TRPV6, a non-voltage gated calcium channel associated with malignancy and poor prognosis in breast cancer.
  • High-affinity TRPV6 antagonist with an IC50 value of 14 nM
  • Demonstrates anticancer activity
  • Targets TRPV6, a non-voltage gated calcium channel
  • Effectively inhibits tumor growth in relevant animal models
Catalog No. 50-004-22646 Supplier Medchemexpress LLC Supplier No. HYP165110MG
SOR-C13 is a carboxy-terminal truncated peptide that acts as a high-affinity TRPV6 antagonist with an IC50 value of 14 nM. It exhibits anticancer activity by targeting TRPV6, a non-voltage gated calcium channel associated with malignancy and poor prognosis in breast cancer.
  • High-affinity TRPV6 antagonist with an IC50 value of 14 nM
  • Demonstrates anticancer activity
  • Targets TRPV6, a non-voltage gated calcium channel
  • Effectively inhibits tumor growth in relevant animal models