Learn More
Medchemexpress LLC SOR-C13 TFA | 1187852-48-7 | 99.3% | 1565.81 (free acid) | 50 MG
SDP
Catalog No. 5000423590
Encompass_Preferred
SOR-C13 TFA is a carboxy-terminal truncated peptide that acts as a high-affinity TRPV6 antagonist, exhibiting an IC50 value of 14 nM. TRPV6 is a non-voltage gated calcium channel implicated in malignancy and poor prognosis in breast cancer. This compound has demonstrated anticancer activity.
  • High-affinity TRPV6 antagonist with an IC50 of 14 nM.
  • TRPV6 is associated with malignancy and poor prognosis in breast cancer.
  • Exhibits anticancer activity.
  • In vivo studies showed effective inhibition of tumor growth in female NOD/SCID mice with SKOV-3 cells at dosages of 400, 600, and 800 mg/kg (IP; daily; on days 1 to 12).
  • Has been investigated in clinical trials for cancer and advanced malignant solid neoplasm.
Catalog No. 50-004-23590 Supplier Medchemexpress LLC Supplier No. HYP1651A50MG
SOR-C13 TFA is a carboxy-terminal truncated peptide that acts as a high-affinity TRPV6 antagonist, exhibiting an IC50 value of 14 nM. TRPV6 is a non-voltage gated calcium channel implicated in malignancy and poor prognosis in breast cancer. This compound has demonstrated anticancer activity.
  • High-affinity TRPV6 antagonist with an IC50 of 14 nM.
  • TRPV6 is associated with malignancy and poor prognosis in breast cancer.
  • Exhibits anticancer activity.
  • In vivo studies showed effective inhibition of tumor growth in female NOD/SCID mice with SKOV-3 cells at dosages of 400, 600, and 800 mg/kg (IP; daily; on days 1 to 12).
  • Has been investigated in clinical trials for cancer and advanced malignant solid neoplasm.