Learn More
Medchemexpress LLC SOR-C13 TFA | 99.32% | 1565.81 (free acid) | 100 MG
SDP
Catalog No. 5000423169
Encompass_Preferred
SOR-C13 TFA is a carboxy-terminal truncated peptide and a high-affinity TRPV6 antagonist with an IC50 value of 14 nM. TRPV6 is a non-voltage gated calcium channel linked to malignancy and poor prognosis in breast cancer. This product exhibits anticancer activity.
  • Solid appearance
  • White to off-white color
  • Lys-Glu-Phe-Leu-His-Pro-Ser-Lys-Val-Asp-Leu-Pro-Arg sequence
  • Soluble in H2O and DMSO (100 mg/mL, requires ultrasonic)
Catalog No. 50-004-23169 Supplier Medchemexpress LLC Supplier No. HYP1651A100MG
SOR-C13 TFA is a carboxy-terminal truncated peptide and a high-affinity TRPV6 antagonist with an IC50 value of 14 nM. TRPV6 is a non-voltage gated calcium channel linked to malignancy and poor prognosis in breast cancer. This product exhibits anticancer activity.
  • Solid appearance
  • White to off-white color
  • Lys-Glu-Phe-Leu-His-Pro-Ser-Lys-Val-Asp-Leu-Pro-Arg sequence
  • Soluble in H2O and DMSO (100 mg/mL, requires ultrasonic)