Learn More
Medchemexpress LLC Sorafenib-13C,d3 | 1210608-86-8 | 98.4% | 468.84 | C20(13C)H13D3ClF3N4O3 | 100 UG
SDP
Catalog No. 5000247425
Encompass_Preferred
Isotopically labeled sorafenib (13C and d3) intended as an analytical standard and internal standard for mass spectrometry, quantitation, and tracer studies. Supplied as a high-purity research reagent for method development and pharmacokinetic sample analysis. For research use only; not for human or clinical use.
  • Isotopically labeled with 13C and three deuteriums.
  • Suitable as an internal standard for LC-MS/MS assays.
  • High purity suitable for quantitative analysis.
  • Supplied in small vials for low-level calibration, e.g. 100 μg.
  • Includes supporting documents such as COA and LC-MS data.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-47425 Supplier Medchemexpress LLC Supplier No. HY10201S2100UG
Isotopically labeled sorafenib (13C and d3) intended as an analytical standard and internal standard for mass spectrometry, quantitation, and tracer studies. Supplied as a high-purity research reagent for method development and pharmacokinetic sample analysis. For research use only; not for human or clinical use.
  • Isotopically labeled with 13C and three deuteriums.
  • Suitable as an internal standard for LC-MS/MS assays.
  • High purity suitable for quantitative analysis.
  • Supplied in small vials for low-level calibration, e.g. 100 μg.
  • Includes supporting documents such as COA and LC-MS data.
  • Intended for research use only; not for clinical or human use.