Learn More
Medchemexpress LLC Sorafenib-13C,d3 | 1210608-86-8 | 98.4% | 468.84 | C2013CH13D3ClF3N4O3 | 10 MG
SDP
Catalog No. 5000247426
Encompass_Preferred
Sorafenib-13C,d3 is a stable isotope-labeled reference standard of sorafenib designed for research and analytical applications. It contains 13C and deuterium labels to provide a defined mass shift for use in LC-MS quantification, tracer studies, and pharmacokinetic experiments. The material is supplied at high purity suitable for use as an internal standard in analytical workflows.
  • High isotopic labeling for reliable mass-spectrometry traceability.
  • High purity suitable for quantitative analysis.
  • Provided in small mg-scale packaging for analytical workflows.
  • Intended for use as an internal standard, tracer, or reference material.
  • Consistent mass shift enables accurate quantitation in LC-MS methods.
Catalog No. 50-002-47426 Supplier Medchemexpress LLC Supplier No. HY10201S210MG
Sorafenib-13C,d3 is a stable isotope-labeled reference standard of sorafenib designed for research and analytical applications. It contains 13C and deuterium labels to provide a defined mass shift for use in LC-MS quantification, tracer studies, and pharmacokinetic experiments. The material is supplied at high purity suitable for use as an internal standard in analytical workflows.
  • High isotopic labeling for reliable mass-spectrometry traceability.
  • High purity suitable for quantitative analysis.
  • Provided in small mg-scale packaging for analytical workflows.
  • Intended for use as an internal standard, tracer, or reference material.
  • Consistent mass shift enables accurate quantitation in LC-MS methods.