Learn More
Medchemexpress LLC Sorafenib-13C,d3 | 1210608-86-8 | 98.4% | 468.84 g·mol⁻¹ | C2013CH13D3ClF3N4O3 | 1 MG
SDP
Catalog No. 5000247352
Encompass_Preferred
Sorafenib-13C,d3 is a stable isotope-labeled analogue of the multikinase inhibitor sorafenib, incorporating 13C and three deuterium atoms for use in analytical and tracer research. It is supplied as a high-purity research reagent for laboratory methods such as LC-MS quantitation and metabolic profiling; not for human or veterinary use.
  • Stable isotope labeling (13C and d3) for tracer and mass spectrometry studies.
  • High purity (98.4%) suitable for analytical applications.
  • Molecular weight 468.84 g·mol⁻¹.
  • Optimized for LC-MS quantitation and metabolic profiling.
  • Available in small, research-scale quantities (e.g., 1 mg vials).
  • For research use only; not intended for clinical use.
Catalog No. 50-002-47352 Supplier Medchemexpress LLC Supplier No. HY10201S21MG
Sorafenib-13C,d3 is a stable isotope-labeled analogue of the multikinase inhibitor sorafenib, incorporating 13C and three deuterium atoms for use in analytical and tracer research. It is supplied as a high-purity research reagent for laboratory methods such as LC-MS quantitation and metabolic profiling; not for human or veterinary use.
  • Stable isotope labeling (13C and d3) for tracer and mass spectrometry studies.
  • High purity (98.4%) suitable for analytical applications.
  • Molecular weight 468.84 g·mol⁻¹.
  • Optimized for LC-MS quantitation and metabolic profiling.
  • Available in small, research-scale quantities (e.g., 1 mg vials).
  • For research use only; not intended for clinical use.