Learn More
Selleck Chemical LLC Sorafenib Tosylate 50mg 475207-59-1 Bay 43-9006
SDP
Catalog No. 506220
Encompass_Preferred
Sorafenib Tosylate (Bay 43-9006) is a multikinase inhibitor of Raf-1 and B-Raf with IC50 of 6 nM and 22 nM in cell-free assays, respectively. Sorafenib Tosylate inhibits VEGFR-2, VEGFR-3, PDGFR-?, Flt-3 and c-KIT with IC50 of 90 nM, 20 nM, 57 nM, 59 nM and 68 nM, respectively. Sorafenib Tosylate induces autophagy and apoptosis and activates ferroptosis with anti-tumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-622-0 Supplier Selleck Chemical LLC Supplier No. S104050MG
Sorafenib Tosylate (Bay 43-9006) is a multikinase inhibitor of Raf-1 and B-Raf with IC50 of 6 nM and 22 nM in cell-free assays, respectively. Sorafenib Tosylate inhibits VEGFR-2, VEGFR-3, PDGFR-?, Flt-3 and c-KIT with IC50 of 90 nM, 20 nM, 57 nM, 59 nM and 68 nM, respectively. Sorafenib Tosylate induces autophagy and apoptosis and activates ferroptosis with anti-tumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.