Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sorafenib-d3 (Donafenib) | 1130115-44-4 | MFCD08460931 | 99.6% | 467.84 g/mol | C21H13D3ClF3N4O3 | 1 ML
SDP

Catalog No. 5000247351
Encompass_Preferred

Sorafenib-d3 is the deuterium-labeled analogue of the multikinase inhibitor sorafenib, supplied as a ready-to-use 10 mM solution in DMSO for research applications, including use as an analytical standard and in pharmacological studies.

  • Deuterium-labeled compound for use as an internal standard.
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.6%) suitable for analytical use.
  • Suitable for biochemical and pharmacological research.
  • Molecular formula C21H13D3ClF3N4O3; molecular weight 467.84 g/mol.

Catalog No. 50-002-47351 Supplier Medchemexpress LLC Supplier No. HY10201S10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sorafenib-d3 is the deuterium-labeled analogue of the multikinase inhibitor sorafenib, supplied as a ready-to-use 10 mM solution in DMSO for research applications, including use as an analytical standard and in pharmacological studies.

  • Deuterium-labeled compound for use as an internal standard.
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.6%) suitable for analytical use.
  • Suitable for biochemical and pharmacological research.
  • Molecular formula C21H13D3ClF3N4O3; molecular weight 467.84 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.