Learn More
Cayman Chemical Sorafenib N-oxide, Purity: ≥98%, CAS: 583840-03-3, 5 mg
SDP
Catalog No. 502940084
Encompass_Preferred
An active metabolite of sorafenib; inhibits FLT3-ITD (Kd = 70 nM); inhibits proliferation of MV4-11 AML cells expressing FLT3-ITD (IC50 = 25.8 nM); selective for AML cell lines containing FLT3-ITD over lines containing wild-type FLT3 (IC50s = 3.9-13.3 µM); a linear-mixed inhibitor of CYP3A4 (Ki = 15 µM)
Catalog No. 50-294-0084 Supplier Cayman Chemical Supplier No. 278305 MG
An active metabolite of sorafenib; inhibits FLT3-ITD (Kd = 70 nM); inhibits proliferation of MV4-11 AML cells expressing FLT3-ITD (IC50 = 25.8 nM); selective for AML cell lines containing FLT3-ITD over lines containing wild-type FLT3 (IC50s = 3.9-13.3 µM); a linear-mixed inhibitor of CYP3A4 (Ki = 15 µM)