Learn More
Apexbio Technology LLC Sorafenib Tosylate 475207-59-1 200mg
SDP
Catalog No. 5000088525
Encompass_Preferred
Sorafenib tosylate (CAS 475207-59-1) is an orally bioavailable multi-kinase inhibitor that targets Raf serine/threonine kinases and vascular endothelial growth factor receptors (VEGFR) thereby impeding tumor cell proliferation and angiogenesis In vitro it demonstrates an IC50 of 2 09 g/ml in HepG2 hepatocellular carcinoma cells and 0 88 nM in MV4-11 (FLT3-ITD) leukemia cells with dose-dependent antiproliferative and pro-apoptotic effects observed in leukemia cell lines at nanomolar concentrations In vivo administration in a breast cancer bone metastasis model significantly reduces osteolytic lesion volume and soft tissue components compared to controls Sorafenib tosylate is widely employed in oncology research to investigate signal transduction tumor growth and angiogenic pathways
Catalog No. 50-000-88525 Supplier Apexbio Technology LLC Supplier No. A8245200
Sorafenib tosylate (CAS 475207-59-1) is an orally bioavailable multi-kinase inhibitor that targets Raf serine/threonine kinases and vascular endothelial growth factor receptors (VEGFR) thereby impeding tumor cell proliferation and angiogenesis In vitro it demonstrates an IC50 of 2 09 g/ml in HepG2 hepatocellular carcinoma cells and 0 88 nM in MV4-11 (FLT3-ITD) leukemia cells with dose-dependent antiproliferative and pro-apoptotic effects observed in leukemia cell lines at nanomolar concentrations In vivo administration in a breast cancer bone metastasis model significantly reduces osteolytic lesion volume and soft tissue components compared to controls Sorafenib tosylate is widely employed in oncology research to investigate signal transduction tumor growth and angiogenic pathways