Learn More
Cayman Chemical Sorafenib (tosylate), Purity: ≥98%, CAS: 475207-59-1, 25 g
SDP
Catalog No. 502913448
Encompass_Preferred
A multi-kinase inhibitor; inhibits Raf-1 and B-RAF (IC50s = 6 and 22 µM, respectively); inhibits VEGFR2, VEGFR3, PDGFRβ, FLT3, and c-Kit (IC50s = 90, 15, 20, 57, and 58 nM, respectively); selective for these kinases over 12 other kinases (IC50s = >10 µM for all); inhibits proliferation of PLC/PRF/5 and HepG2 cells (IC50s = 6.3 and 4.5 µM, respectively); inhibits tumor growth in a PLC/PRF/5 mouse xenograft model at 30 mg/kg; induces ferroptotic cell death in HT-1080 fibrosarcoma cells at 10 µM but not in a variety of other cancer cell lines; inhibits HCV replication in Huh7.5 cells (IC50 = 7.2 µM)
Catalog No. 50-291-3448 Supplier Cayman Chemical Supplier No. 3561225 G
A multi-kinase inhibitor; inhibits Raf-1 and B-RAF (IC50s = 6 and 22 µM, respectively); inhibits VEGFR2, VEGFR3, PDGFRβ, FLT3, and c-Kit (IC50s = 90, 15, 20, 57, and 58 nM, respectively); selective for these kinases over 12 other kinases (IC50s = >10 µM for all); inhibits proliferation of PLC/PRF/5 and HepG2 cells (IC50s = 6.3 and 4.5 µM, respectively); inhibits tumor growth in a PLC/PRF/5 mouse xenograft model at 30 mg/kg; induces ferroptotic cell death in HT-1080 fibrosarcoma cells at 10 µM but not in a variety of other cancer cell lines; inhibits HCV replication in Huh7.5 cells (IC50 = 7.2 µM)