Learn More
Medchemexpress LLC HY-100414 1mg Medchemexpress, Soraprazan CAS:261944-46-1 Purity:>98%
SDP
Catalog No. 501931821
Encompass_Preferred
Medchemexpress, HY-100414 1mg Soraprazan CAS:261944-46-1 Soraprazan (BYK61359) is a reversible, and fast-acting inhibitor of gastric H + /K + ATPase . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1821 Supplier Medchemexpress LLC Supplier No. HY1004141MG
Medchemexpress, HY-100414 1mg Soraprazan CAS:261944-46-1 Soraprazan (BYK61359) is a reversible, and fast-acting inhibitor of gastric H + /K + ATPase . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.